Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198526.3 | NP_940928.2 | 3 | 1764 | ||
XM_005254906.3 | XP_005254963.1 | 3 | 1790 | |||
XM_005254907.3 | XP_005254964.1 | 3 | 1711 | |||
XM_011521540.2 | XP_011519842.1 | 4 | 1872 | |||
XM_011521541.2 | XP_011519843.1 | 4 | 1833 | |||
XM_011521542.2 | XP_011519844.1 | 3 | 1751 | |||
XM_011521543.2 | XP_011519845.1 | 2 | 1640 | |||
XM_011521544.1 | XP_011519846.1 | 2 | 1640 | |||
GenBank mRNA | AK094712.1 | - | 3 | 1735 | ||
AK160373.1 | - | 2 | 454 | |||
AL831826.1 | - | 2 | 235 | |||
BC126476.1 | - | 2 | 1636 | 8992192 | ||
BC130541.1 | - | 2 | 1636 | 40146572 |