Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198529.3 | NP_940931.2 | 16 | 3337 | ||
XM_011524759.1 | XP_011523061.1 | 16 | 3343 | |||
XM_011524763.2 | XP_011523065.1 | 16 | 3343 | |||
XM_011524765.1 | XP_011523067.1 | 16 | 3343 | |||
XM_011524766.2 | XP_011523068.1 | 11 | 2283 | |||
XM_017024591.1 | XP_016880080.1 | 13 | 2422 | |||
GenBank mRNA | AK128126.1 | - | 11 | 2191 | ||
BC110608.1 | - | 13 | 2540 | |||
BC110609.1 | - | 13 | 2540 | |||
BC137519.1 | - | 16 | 3173 | 9021519 |