Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRC37A3 SIRNA Assays ›
Gene Symbol:
LRRC37A3
Gene Name:
leucine rich repeat containing 37 member A3
Gene Aliases:
LRRC37, LRRC37A
Chromosome Location:
Chr.17: 64854147 - 64966228 on Build GRCh38
UniGene ID:
Hs.551962
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s51587
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRC37A3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303255.1
NP_001290184.1
8
802
NM_199340.3
NP_955372.2
12
4059
XM_011524776.1
XP_011523078.1
9
1498
XM_011524777.1
XP_011523079.1
7
1345
XM_011524778.2
XP_011523080.2
10
1023
XM_017024593.1
XP_016880082.1
10
3888
XM_017024594.1
XP_016880083.1
9
3816
XM_017024595.1
XP_016880084.1
9
3816
XM_017024596.1
XP_016880085.1
8
3735
XM_017024597.1
XP_016880086.1
8
3735
XM_017024598.1
XP_016880087.1
7
3663
XM_017024599.1
XP_016880088.1
11
3260
XM_017024601.1
XP_016880090.1
9
1200
XM_017024602.1
XP_016880091.1
6
975
XM_017024603.1
XP_016880092.1
9
1498
XM_017024604.1
XP_016880093.1
8
1426
XM_017024605.1
XP_016880094.1
7
1345
XM_017024606.1
XP_016880095.1
6
1273
XM_017024607.1
XP_016880096.1
9
803
XM_017024608.1
XP_016880097.1
9
1820
GenBank mRNA
AB011135.2
-
11
3924
AK091625.1
-
2
907
AK294371.1
-
8
790
AK299714.1
-
5
556
AK308554.1
-
5
535
AK315945.1
-
9
1531
BC043145.1
-
9
3401
5296147
BC113643.1
-
2
421
BC117274.1
-
2
421
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline