Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP9B SIRNA Assays ›
Gene Symbol:
ATP9B
Gene Name:
ATPase phospholipid transporting 9B (putative)
Gene Aliases:
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
Chromosome Location:
Chr.18: 79069275 - 79378391 on Build GRCh38
UniGene ID:
Hs.465475
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s51593
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP9B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306085.1
NP_001293014.1
9
1065
NM_198531.4
NP_940933.3
9
1065
XM_011525963.2
XP_011524265.1
10
1231
XM_011525964.2
XP_011524266.1
9
946
XM_011525965.2
XP_011524267.1
10
1227
XM_011525966.2
XP_011524268.1
9
1182
XM_011525971.2
XP_011524273.2
3
3289
XM_011525972.2
XP_011524274.1
4
306
XM_011525973.2
XP_011524275.1
2
80
XM_011525974.2
XP_011524276.1
2
102
XM_017025726.1
XP_016881215.1
10
1231
XM_017025727.1
XP_016881216.1
10
1231
XM_017025728.1
XP_016881217.1
10
1231
XM_017025729.1
XP_016881218.1
10
1231
XM_017025730.1
XP_016881219.1
10
1231
XM_017025731.1
XP_016881220.1
10
1231
XM_017025732.1
XP_016881221.1
10
1231
XM_017025733.1
XP_016881222.1
10
1231
XM_017025734.1
XP_016881223.1
10
1231
XM_017025735.1
XP_016881224.1
10
1231
XM_017025736.1
XP_016881225.1
10
1231
XM_017025737.1
XP_016881226.1
10
1231
XM_017025738.1
XP_016881227.1
6
541
XM_017025739.1
XP_016881228.1
7
636
XM_017025740.1
XP_016881229.1
6
541
XM_017025741.1
XP_016881230.1
6
541
XM_017025742.1
XP_016881231.1
10
1231
GenBank mRNA
AB373110.1
-
8
773
AK093704.1
-
7
665
AK097757.1
-
8
1087
AK308291.1
-
9
947
BC125219.1
-
9
943
40123958
BC125220.1
-
9
943
40123959
BX537860.1
-
7
448
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
retrograde vesicle-mediated transport, Golgi to ER
endocytosis
phospholipid translocation
establishment of protein localization to Golgi
magnesium ion binding
phospholipid-translocating ATPase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline