Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320865.1 | NP_001307794.1 | 7 | 717 | ||
NM_198559.1 | NP_940961.1 | 7 | 684 | |||
XM_005246539.4 | XP_005246596.1 | 6 | 681 | |||
XM_005246541.4 | XP_005246598.1 | 5 | 480 | |||
XM_011511148.2 | XP_011509450.1 | 6 | 648 | |||
XM_011511149.2 | XP_011509451.1 | 5 | 479 | |||
XM_011511150.2 | XP_011509452.1 | 5 | 445 | |||
XM_017004053.1 | XP_016859542.1 | 5 | 880 | |||
GenBank mRNA | BC052750.1 | - | 7 | 684 | 5742766 |