Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135914.1 | NP_001129386.1 | 13 | 1358 | ||
NM_199349.2 | NP_955381.2 | 15 | 1532 | |||
XM_017012184.1 | XP_016867673.1 | 15 | 1532 | |||
XM_017012185.1 | XP_016867674.1 | 15 | 1532 | |||
XM_017012186.1 | XP_016867675.1 | 15 | 1532 | |||
XM_017012187.1 | XP_016867676.1 | 15 | 1532 | |||
XM_017012188.1 | XP_016867677.1 | 15 | 1532 | |||
XM_017012189.1 | XP_016867678.1 | 15 | 1532 | |||
XM_017012190.1 | XP_016867679.1 | 15 | 1532 | |||
XM_017012191.1 | XP_016867680.1 | 8 | 751 | |||
XM_017012192.1 | XP_016867681.1 | 8 | 762 | |||
XM_017012193.1 | XP_016867682.1 | 15 | 1532 | |||
XM_017012194.1 | XP_016867683.1 | 15 | 1532 | |||
GenBank mRNA | AK095464.1 | - | 8 | 1006 | ||
AK122706.1 | - | 15 | 1532 |