Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010895.2 | NP_001010895.1 | 2 | 821 | ||
NM_020207.4 | NP_064592.2 | 2 | 821 | |||
XM_011518641.2 | XP_011516943.1 | 2 | 821 | |||
XM_011518644.2 | XP_011516946.1 | 2 | 807 | |||
XM_011518645.2 | XP_011516947.1 | 2 | 821 | |||
XM_011518646.2 | XP_011516948.1 | 2 | 821 | |||
XM_011518647.1 | XP_011516949.1 | 2 | 821 | |||
XM_017014707.1 | XP_016870196.1 | 2 | 807 | |||
GenBank mRNA | AK307504.1 | - | 2 | 467 | ||
AK308409.1 | - | 2 | 753 | |||
AL389953.1 | - | 2 | 737 | |||
BC035183.1 | - | 2 | 1322 | 5266310 | ||
BC075860.1 | - | 1 | 62 | |||
BC140702.1 | - | 2 | 564 | 9021570 | ||
BC144261.1 | - | 2 | 564 |