Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308154.1 | NP_001295083.1 | 1, 2 | 208 | ||
NM_198686.2 | NP_941959.1 | 1, 2 | 208 | |||
GenBank mRNA | AB672631.2 | - | 1, 2 | 177 | ||
AB672632.1 | - | 1, 2 | 178 | |||
AB678452.1 | - | 1, 2 | 208 | |||
AB678453.1 | - | 1, 2 | 191 | |||
BC040679.2 | - | 1, 2 | 131 | 4817835 | ||
CN352234.1 | - | 1, 2 | 209 |