Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198581.2 | NP_940983.2 | 8 | 1445 | ||
XM_006712519.2 | XP_006712582.1 | 8 | 1452 | |||
GenBank mRNA | AB111887.1 | - | 7 | 891 | ||
AK127776.1 | - | 5 | 514 | |||
AK131416.1 | - | 8 | 1431 | |||
BC155540.1 | - | 7 | 891 | |||
BC167154.1 | - | 7 | 891 |