Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032450.2 | NP_115826.2 | 23, 24 | 2570 | ||
XM_011517270.2 | XP_011515572.1 | 23, 24 | 2708 | |||
XM_011517271.2 | XP_011515573.1 | 23, 24 | 2707 | |||
XM_011517273.2 | XP_011515575.1 | 23, 24 | 2706 | |||
XM_011517275.2 | XP_011515577.1 | 23, 24 | 2706 | |||
XM_011517276.2 | XP_011515578.1 | 23, 24 | 2734 | |||
XM_011517277.1 | XP_011515579.1 | 23, 24 | 2570 | |||
XM_011517278.1 | XP_011515580.1 | 22, 23 | 2450 | |||
XM_011517279.2 | XP_011515581.1 | 22, 23 | 2614 | |||
XM_011517281.1 | XP_011515583.1 | 22, 23 | 2623 | |||
XM_011517282.1 | XP_011515584.1 | 21, 22 | 2548 | |||
XM_011517283.2 | XP_011515585.1 | 23, 24 | 2529 | |||
XM_017013806.1 | XP_016869295.1 | 20, 21 | 2185 | |||
GenBank mRNA | AL834309.1 | - | 22, 23 | 2454 |