Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012279.2 | NP_001012279.1 | 6 | 2480 | ||
GenBank mRNA | AK091416.1 | - | 3 | 251 | ||
AK096490.1 | - | 5 | 925 | |||
BC127760.1 | - | 5 | 749 | |||
BC143043.1 | - | 5 | 749 | |||
BC143044.1 | - | 5 | 749 |