Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004467.3 | NP_004458.3 | 6 | 639 | ||
NM_147203.2 | NP_671736.2 | 6 | 727 | |||
NM_201552.1 | NP_963846.1 | 7 | 766 | |||
NM_201553.1 | NP_963847.1 | 7 | 854 | |||
XM_017013232.1 | XP_016868721.1 | 7 | 1616 | |||
XM_017013233.1 | XP_016868722.1 | 5 | 691 | |||
GenBank mRNA | AF042354.1 | - | 6 | 675 | ||
AF168954.1 | - | 6 | 662 | |||
AK225616.1 | - | 6 | 625 | |||
AK225619.1 | - | 6 | 614 | |||
AK289448.1 | - | 6 | 625 | |||
AX772838.1 | - | 7 | 719 | |||
BC007047.1 | - | 6 | 635 | 4071810 | ||
BT006635.1 | - | 5 | 530 | |||
D14446.1 | - | 6 | 608 | |||
D87342.1 | - | 5 | 535 |