Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162505.1 | NP_001155977.1 | 6 | 1811 | ||
NM_199129.2 | NP_954580.1 | 6 | 1820 | |||
NR_027889.1 | 7 | 1924 | ||||
GenBank mRNA | AB044550.1 | - | 6 | 1733 | ||
AF155120.1 | - | 6 | 1747 | |||
AK023028.1 | - | 6 | 1745 | |||
BC018893.2 | - | 6 | 1731 | |||
BC142966.1 | - | 6 | 1733 | 8860386 | ||
BC144612.1 | - | 7 | 1925 |