Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199352.3 | NP_955384.3 | 8 | 1374 | ||
XM_017017684.1 | XP_016873173.1 | 12 | 2543 | |||
XM_017017685.1 | XP_016873174.1 | 13 | 2671 | |||
XM_017017686.1 | XP_016873175.1 | 12 | 2979 | |||
XM_017017687.1 | XP_016873176.1 | 11 | 2994 | |||
XM_017017688.1 | XP_016873177.1 | 8 | 1932 | |||
XM_017017689.1 | XP_016873178.1 | 9 | 1844 | |||
XM_017017690.1 | XP_016873179.1 | 11 | 1747 | |||
XM_017017691.1 | XP_016873180.1 | 9 | 1200 | |||
XM_017017692.1 | XP_016873181.1 | 8 | 1371 | |||
XM_017017693.1 | XP_016873182.1 | 7 | 962 | |||
XM_017017694.1 | XP_016873183.1 | 7 | 1801 | |||
XM_017017696.1 | XP_016873185.1 | 9 | 1503 | |||
GenBank mRNA | AY437532.1 | - | 8 | 1374 | ||
BC101315.1 | - | 9 | 1521 | |||
BC101316.1 | - | 8 | 1374 | |||
BC101317.1 | - | 6 | 1098 |