Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMS1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003611.2 | 2 | 237 | |||
GenBank mRNA | AK307333.1 | - | 2 | 96 | ||
AL832069.1 | - | 2 | 124 | |||
BC017177.2 | - | 1 | 85 | |||
BC065722.1 | - | 2 | 106 | |||
BC131568.1 | - | 1 | 891 | |||
DA245014.1 | - | 2 | 130 |
Gene Symbol: BMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014753.3 | NP_055568.3 | 2 | 399 | ||
XM_005271846.3 | XP_005271903.1 | 2 | 274 | |||
XM_005271848.3 | XP_005271905.1 | 2 | 130 | |||
XM_005271849.3 | XP_005271906.1 | 2 | 130 | |||
XM_011540402.2 | XP_011538704.1 | 2 | 130 | |||
GenBank mRNA | AK308420.1 | - | 2 | 106 | ||
BC043345.1 | - | 2 | 94 | 6062924 | ||
BC150252.1 | - | 1 | 68 | 9007274 | ||
BX485446.1 | - | 2 | 405 | |||
D80009.1 | - | 1 | 68 | |||
DB064246.1 | - | 2 | 368 |
Gene Symbol: BMS1P21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033857.1 | 2 | 226 | |||
GenBank mRNA | AK307851.1 | - | 2 | 218 |