Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213606.3 | NP_998771.3 | 4 | 555 | ||
XM_017016237.1 | XP_016871726.1 | 3 | 645 | |||
XM_017016238.1 | XP_016871727.1 | 3 | 584 | |||
XM_017016239.1 | XP_016871728.1 | 3 | 443 | |||
GenBank mRNA | AK127303.1 | - | 4 | 556 | ||
CR936657.1 | - | 3 | 414 |