Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001732.3 | NP_001001732.2 | 6 | 626 | ||
NM_001159747.1 | NP_001153219.1 | 7 | 659 | |||
XM_011539789.2 | XP_011538091.1 | 14 | 1720 | |||
XM_011539791.2 | XP_011538093.1 | 13 | 1537 | |||
XM_011539792.2 | XP_011538094.1 | 8 | 842 | |||
XM_011539793.2 | XP_011538095.1 | 7 | 1821 | |||
XM_011539795.2 | XP_011538097.1 | 6 | 1638 | |||
XM_017016240.1 | XP_016871729.1 | 7 | 898 | |||
GenBank mRNA | AK302377.1 | - | 7 | 638 | ||
BC075861.1 | - | 6 | 591 | 5171414 |