Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129998.2 | NP_001123470.1 | 4 | 602 | ||
NM_001319241.1 | NP_001306170.1 | 4 | 594 | |||
NM_001319242.1 | NP_001306171.1 | 4 | 594 | |||
NM_205852.3 | NP_995324.2 | 4 | 602 | |||
NR_135049.1 | 4 | 602 | ||||
XM_011520658.2 | XP_011518960.1 | 5 | 613 | |||
XM_017019295.1 | XP_016874784.1 | 5 | 674 | |||
GenBank mRNA | AK128243.1 | - | 4 | 597 | ||
AY358810.1 | - | 4 | 602 | |||
BC036036.1 | - | 4 | 462 | |||
BC136805.1 | - | 4 | 542 | 9020795 | ||
BC136806.1 | - | 4 | 542 | 9020796 | ||
BM563861.1 | - | 461 | ||||
DQ368812.1 | - | 4 | 469 |