Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039492.2 | NP_001034581.1 | 4 | 493 | ||
NM_001318894.1 | NP_001305823.1 | 3 | 739 | |||
NM_001318895.1 | NP_001305824.1 | 4 | 727 | |||
NM_001318896.1 | NP_001305825.1 | 3 | 442 | |||
NM_001318899.1 | NP_001305828.1 | 5 | 615 | |||
NM_001450.3 | NP_001441.4 | 3 | 676 | |||
NM_201555.1 | NP_963849.1 | 5 | 848 | |||
NM_201557.3 | NP_963851.2 | 3 | 668 | |||
XM_011510798.2 | XP_011509100.1 | 4 | 475 | |||
GenBank mRNA | AK313841.1 | - | 4 | 525 | ||
AL521653.3 | - | 3 | 369 | |||
AY634685.1 | - | 2 | 269 | |||
BC012742.2 | - | 5 | 848 | 3840517 | ||
BC014397.1 | - | 5 | 848 | 3546686 | ||
BC093049.1 | - | 3 | 434 | 30374919 | ||
BM559077.1 | - | 5 | 497 | |||
BT006960.1 | - | 2 | 269 | |||
CF552311.1 | - | 3 | 399 | |||
CR457437.1 | - | 2 | 269 | |||
CR936682.1 | - | 3 | 739 | |||
CR936799.1 | - | 3 | 444 | |||
DQ266432.1 | - | 4 | 727 | |||
DQ307067.1 | - | 3 | 606 | |||
JQ313902.1 | - | 2 | 239 | |||
L42176.1 | - | 4 | 407 | |||
U29332.1 | - | 3 | 390 |