Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001436.2 | NP_001001436.1 | 3 | 235 | ||
GenBank mRNA | BC056676.1 | - | 3 | 239 | 6711353 |