Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014985.2 | NP_001014985.2 | 1 | 108 | ||
XM_005256626.3 | XP_005256683.1 | 1 | 151 | |||
GenBank mRNA | BC029290.1 | - | 1 | 108 |