Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167985.1 | NP_001161457.1 | 6 | 499 | ||
NM_001167986.1 | NP_001161458.1 | 7 | 831 | |||
NM_001167987.1 | NP_001161459.1 | 6 | 544 | |||
NM_213726.2 | NP_998891.2 | 6 | 661 | |||
XM_005256627.3 | XP_005256684.1 | 5 | 739 | |||
XM_005256628.4 | XP_005256685.1 | 5 | 936 | |||
XM_005256629.1 | XP_005256686.1 | 5 | 746 | |||
XM_006721517.1 | XP_006721580.1 | 5 | 791 | |||
XM_006721518.2 | XP_006721581.1 | 5 | 695 | |||
XM_006721519.3 | XP_006721582.1 | 5 | 892 | |||
XM_011523832.2 | XP_011522134.1 | 6 | 1061 | |||
XM_011523834.2 | XP_011522136.1 | 5 | 739 | |||
GenBank mRNA | AY601906.1 | - | 6 | 661 | ||
AY601907.1 | - | 6 | 499 | |||
AY604176.1 | - | 4 | 219 | |||
BC119781.1 | - | 6 | 620 | 40109268 | ||
BC122873.1 | - | 5 | 333 | 40109270 |