Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000972.2 | NP_000963.1 | 6, 7 | 654 | ||
GenBank mRNA | AB056665.1 | - | 3, 4 | 253 | ||
AK222534.1 | - | 6, 7 | 651 | |||
AK291123.1 | - | 6, 7 | 646 | |||
AK311743.1 | - | 6, 7 | 650 | |||
BC005128.1 | - | 1 | 637 | 3938260 | ||
BC021052.1 | - | 1 | 628 | |||
BC021979.2 | - | 1 | 626 | 4053567 | ||
BC023594.2 | - | 6, 7 | 685 | 4841982 | ||
BC023624.2 | - | 6, 7 | 690 | 4810336 | ||
BC032533.1 | - | 1 | 673 | |||
BC071900.1 | - | 1 | 626 | 6090366 | ||
BC071901.1 | - | 5, 6 | 634 | 6262332 | ||
BC073802.1 | - | 1 | 625 | 4336858 | ||
BC105290.1 | - | 1 | 651 | 6495031 | ||
M36072.1 | - | 6, 7 | 655 | |||
X06705.1 | - | 6, 7 | 654 |