Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204118.1 | NP_001191047.1 | 10 | 687 | ||
NM_207390.3 | NP_997273.3 | 10 | 687 | |||
NR_109784.1 | 10 | 687 | ||||
NR_109785.1 | 10 | 687 | ||||
XM_017026785.1 | XP_016882274.1 | 11 | 1231 | |||
XM_017026786.1 | XP_016882275.1 | 11 | 1095 | |||
XM_017026787.1 | XP_016882276.1 | 12 | 1450 | |||
XM_017026788.1 | XP_016882277.1 | 11 | 1207 | |||
XM_017026789.1 | XP_016882278.1 | 7 | 909 | |||
XM_017026790.1 | XP_016882279.1 | 10 | 1183 | |||
XM_017026792.1 | XP_016882281.1 | 11 | 1235 | |||
XM_017026793.1 | XP_016882282.1 | 11 | 1238 | |||
XM_017026794.1 | XP_016882283.1 | 11 | 1240 | |||
GenBank mRNA | BC140848.1 | - | 10 | 648 | 9021716 | |
BC144664.1 | - | 10 | 648 | |||
BC144665.1 | - | 10 | 648 | 9053196 |