Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM69A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006605.4 | NP_001006606.2 | 3 | 299 | ||
NM_001252269.1 | NP_001239198.1 | 2 | 164 | |||
NM_001252270.1 | NP_001239199.1 | 3 | 299 | |||
NM_001252271.1 | NP_001239200.1 | 3 | 342 | |||
NM_001252273.1 | NP_001239202.1 | 3 | 299 | |||
GenBank mRNA | AK303700.1 | - | 2 | 317 | ||
AL536469.3 | - | 3 | 243 | |||
AL702320.1 | - | 3 | 263 | |||
BC070342.1 | - | 3 | 233 | 6498824 | ||
BF979329.1 | - | 3 | 243 | |||
DA141589.1 | - | 3 | 312 | |||
DA155419.1 | - | 3 | 216 | |||
DA348631.1 | - | 3 | 342 | |||
DA736826.1 | - | 2 | 40 | |||
DB199113.1 | - | 2 | 90 | |||
DB475309.1 | - | 3 | 299 | |||
DB498496.1 | - | 3 | 442 |