Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM69A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006605.4 | NP_001006606.2 | 3, 4 | 386 | ||
NM_001252269.1 | NP_001239198.1 | 2, 3 | 251 | |||
NM_001252271.1 | NP_001239200.1 | 3, 4 | 429 | |||
NM_001252273.1 | NP_001239202.1 | 3, 4 | 386 | |||
GenBank mRNA | AK303700.1 | - | 2, 3 | 404 | ||
AL536469.3 | - | 3, 4 | 330 | |||
AL702320.1 | - | 3, 4 | 350 | |||
BC070342.1 | - | 3, 4 | 320 | 6498824 | ||
DA141589.1 | - | 3, 4 | 399 | |||
DA155419.1 | - | 3, 4 | 303 | |||
DA348631.1 | - | 3, 4 | 429 | |||
DA736826.1 | - | 2, 3 | 127 | |||
DB199113.1 | - | 2, 3 | 177 | |||
DB475309.1 | - | 3, 4 | 386 |