Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010898.2 | NP_001010898.1 | 10 | 2067 | ||
XM_006710643.2 | XP_006710706.1 | 10 | 1775 | |||
GenBank mRNA | AK289982.1 | - | 10 | 2063 | ||
AK297429.1 | - | 1 | 50 | |||
BC140908.1 | - | 9 | 1652 | 9056720 |