Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 4 | 443 | |||
NR_027791.3 | 2 | 289 | ||||
NR_111004.2 | 3 | 416 | ||||
NR_111005.2 | 3 | 416 | ||||
NR_136541.1 | 4 | 496 | ||||
NR_136542.1 | 4 | 496 | ||||
NR_136543.1 | 2 | 289 | ||||
NR_136551.1 | 2 | 324 | ||||
NR_136552.1 | 2 | 324 | ||||
GenBank mRNA | AK095614.1 | - | 4 | 443 | ||
AL700268.1 | - | 2 | 200 | |||
CF135136.1 | - | 2 | 296 | |||
CF135139.1 | - | 2 | 296 | |||
CN313455.1 | - | 3 | 416 |