Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012186.2 | NP_036318.1 | 1 | 1822 | ||
GenBank mRNA | AF275722.1 | - | 1 | 1833 |