Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161009.2 | NP_001154481.1 | 6 | 1361 | ||
NM_001161010.2 | NP_001154482.1 | 6 | 1289 | |||
NM_001161011.2 | NP_001154483.1 | 5 | 1289 | |||
NM_001161012.2 | NP_001154484.1 | 5 | 1184 | |||
NM_001161013.2 | NP_001154485.1 | 4 | 1112 | |||
NM_031905.4 | NP_114111.2 | 7 | 1466 | |||
XM_011516601.2 | XP_011514903.1 | 6 | 1539 | |||
XM_011516602.2 | XP_011514904.1 | 6 | 900 | |||
XM_017012684.1 | XP_016868173.1 | 8 | 1647 | |||
XM_017012685.1 | XP_016868174.1 | 7 | 1575 | |||
XM_017012686.1 | XP_016868175.1 | 7 | 1470 | |||
XM_017012687.1 | XP_016868176.1 | 6 | 879 | |||
GenBank mRNA | AK225082.1 | - | 6 | 1094 |