Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024613.3 | NP_001019784.2 | 1 | 761 | ||
NM_001160264.2 | NP_001153736.1 | 2 | 611 | |||
XM_005250337.3 | XP_005250394.1 | 2 | 2432 | |||
XM_011516202.2 | XP_011514504.1 | 3 | 2282 | |||
GenBank mRNA | AK300742.1 | - | 1 | 890 | ||
AY726588.1 | - | 1 | 682 | |||
BC127714.1 | - | 1 | 749 | 40127896 | ||
BC127715.1 | - | 2 | 599 | 40127897 | ||
BC136684.1 | - | 1 | 817 | 9020674 | ||
BC144367.1 | - | 1 | 817 | 9052888 |