Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289973.1 | NP_001276902.1 | 8 | 1248 | ||
NM_207411.4 | NP_997294.3 | 7 | 1110 | |||
XM_011534750.2 | XP_011533052.1 | 3 | 1616 | |||
XM_017013401.1 | XP_016868890.1 | 6 | 1529 | |||
XM_017013402.1 | XP_016868891.1 | 5 | 1321 | |||
GenBank mRNA | AY358489.1 | - | 7 | 981 | ||
AY534242.1 | - | 7 | 1110 | |||
AY534243.1 | - | 8 | 1102 |