Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004118.3 | NP_004109.1 | 1 | 220 | ||
GenBank mRNA | AF042831.1 | - | 1 | 121 | ||
BC013408.2 | - | 1 | 220 | 3028182 | ||
BT009768.1 | - | 1 | 145 |