Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000088.3 | NP_000079.2 | 51 | 4646 | ||
XM_005257058.3 | XP_005257115.2 | 49 | 4385 | |||
XM_005257059.4 | XP_005257116.2 | 38 | 3737 | |||
XM_011524341.1 | XP_011522643.1 | 48 | 4457 | |||
GenBank mRNA | AB209597.1 | - | 51 | 4621 | ||
BC036531.2 | - | 51 | 4636 | 5264710 | ||
K01228.1 | - | 33 | 3247 | |||
M32798.1 | - | 1 | 203 | |||
Z74615.1 | - | 51 | 4639 |