Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001396.3 | NP_001387.2 | 6 | 860 | ||
NM_101395.2 | NP_567824.1 | 8 | 1034 | |||
NM_130436.2 | NP_569120.1 | 6 | 2228 | |||
NM_130438.2 | NP_569122.1 | 6 | 861 | |||
XM_005260933.4 | XP_005260990.1 | 2 | 187 | |||
XM_006723976.3 | XP_006724039.1 | 7 | 932 | |||
XM_006723977.3 | XP_006724040.1 | 7 | 1257 | |||
XM_006723978.3 | XP_006724041.1 | 7 | 1986 | |||
XM_006723979.3 | XP_006724042.1 | 7 | 1092 | |||
XM_011529482.2 | XP_011527784.1 | 6 | 954 | |||
XM_011529483.2 | XP_011527785.1 | 7 | 3800 | |||
XM_011529484.2 | XP_011527786.1 | 6 | 927 | |||
XM_011529485.2 | XP_011527787.1 | 6 | 1443 | |||
XM_017028284.1 | XP_016883773.1 | 7 | 891 | |||
XM_017028285.1 | XP_016883774.1 | 7 | 1137 | |||
XM_017028286.1 | XP_016883775.1 | 9 | 1134 | |||
GenBank mRNA | AF108830.1 | - | 8 | 1034 | ||
AK301752.1 | - | 2 | 200 | |||
D85759.1 | - | 6 | 826 | |||
D86550.1 | - | 6 | 2231 | |||
U52373.1 | - | 6 | 860 | |||
U58496.1 | - | 6 | 820 |