Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001396.3 | NP_001387.2 | 5 | 674 | ||
NM_101395.2 | NP_567824.1 | 7 | 848 | |||
NM_130436.2 | NP_569120.1 | 5 | 2042 | |||
NM_130438.2 | NP_569122.1 | 5 | 675 | |||
XM_006723976.3 | XP_006724039.1 | 6 | 746 | |||
XM_006723977.3 | XP_006724040.1 | 6 | 1071 | |||
XM_006723978.3 | XP_006724041.1 | 6 | 1800 | |||
XM_006723979.3 | XP_006724042.1 | 6 | 906 | |||
XM_011529482.2 | XP_011527784.1 | 5 | 768 | |||
XM_011529483.2 | XP_011527785.1 | 6 | 3614 | |||
XM_011529484.2 | XP_011527786.1 | 5 | 741 | |||
XM_011529485.2 | XP_011527787.1 | 5 | 1257 | |||
XM_017028284.1 | XP_016883773.1 | 6 | 705 | |||
XM_017028285.1 | XP_016883774.1 | 6 | 951 | |||
XM_017028286.1 | XP_016883775.1 | 8 | 948 | |||
GenBank mRNA | AF108830.1 | - | 7 | 848 | ||
D85759.1 | - | 5 | 640 | |||
D86550.1 | - | 5 | 2045 | |||
U52373.1 | - | 5 | 674 |