Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040002.1 | NP_001035091.1 | 3 | 499 | ||
NM_004527.3 | NP_004518.1 | 2 | 645 | |||
XM_011524818.1 | XP_011523120.1 | 2 | 1036 | |||
GenBank mRNA | AK291139.1 | - | 2 | 1005 | ||
AW410297.1 | - | 3 | 517 | |||
BC069474.1 | - | 2 | 642 | 7262102 | ||
BC069506.1 | - | 2 | 645 | 7262114 | ||
BE898972.1 | - | 498 | ||||
U10492.1 | - | 2 | 645 |