Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040002.1 | NP_001035091.1 | 2, 3 | 359 | ||
NM_004527.3 | NP_004518.1 | 1, 2 | 505 | |||
XM_011524818.1 | XP_011523120.1 | 1, 2 | 896 | |||
GenBank mRNA | AK291139.1 | - | 1, 2 | 865 | ||
AW410297.1 | - | 2, 3 | 377 | |||
BC069474.1 | - | 1, 2 | 502 | 7262102 | ||
BC069506.1 | - | 1, 2 | 505 | 7262114 | ||
BE898972.1 | - | 359 | ||||
U10492.1 | - | 1, 2 | 505 |