Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032409.2 | NP_001027581.1 | 6 | 1347 | ||
NM_016816.3 | NP_058132.2 | 6 | 1445 | |||
XM_006719434.1 | XP_006719497.1 | 6 | 2064 | |||
XM_011538413.1 | XP_011536715.1 | 6 | 1150 | |||
XM_017019361.1 | XP_016874850.1 | 6 | 1248 | |||
XM_017019362.1 | XP_016874851.1 | 6 | 1097 | |||
GenBank mRNA | AK225384.1 | - | 6 | 1245 | ||
AY730627.1 | - | 6 | 1190 | |||
AY730628.1 | - | 6 | 1093 | |||
DQ914956.1 | - | 1 | 55 | |||
X02661.1 | - | 3 | 491 | |||
X02875.1 | - | 3 | 315 |