Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHEX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000444.5 | NP_000435.3 | 11 | 1811 | ||
NM_001282754.1 | NP_001269683.1 | 11 | 1811 | |||
XM_011545533.1 | XP_011543835.1 | 7 | 620 | |||
XM_011545536.2 | XP_011543838.1 | 3 | 653 | |||
XM_017029579.1 | XP_016885068.1 | 7 | 621 | |||
GenBank mRNA | AD000712.1 | - | 11 | 1408 | ||
AK298030.1 | - | 11 | 1357 | |||
AK301528.1 | - | 10 | 1547 | |||
AK304551.1 | - | 4 | 648 | |||
BC105057.1 | - | 11 | 1333 | 8144060 | ||
BC105059.1 | - | 11 | 1333 | 8144062 | ||
U60475.1 | - | 11 | 1242 | |||
U75645.1 | - | 11 | 1325 | |||
U82970.1 | - | 11 | 1852 | |||
U87284.1 | - | 11 | 1450 |