Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000304.3 | NP_000295.1 | 3 | 404 | ||
NM_001281455.1 | NP_001268384.1 | 3 | 365 | |||
NM_001281456.1 | NP_001268385.1 | 3 | 400 | |||
NM_153321.2 | NP_696996.1 | 3 | 361 | |||
NM_153322.2 | NP_696997.1 | 2 | 283 | |||
NR_104017.1 | 2 | 292 | ||||
XM_017024775.1 | XP_016880264.1 | 2 | 200 | |||
XM_017024776.1 | XP_016880265.1 | 3 | 373 | |||
GenBank mRNA | AK290640.1 | - | 3 | 314 | ||
AK300690.1 | - | 3 | 256 | |||
BC019040.2 | - | 3 | 326 | 4588473 | ||
BC091499.1 | - | 3 | 321 | 30395315 | ||
BF725606.1 | - | 3 | 319 | |||
BQ232542.1 | - | 1 | 73 | |||
BQ694106.1 | - | 3 | 350 | |||
BX464118.2 | - | 3 | 359 | |||
CR541931.1 | - | 2 | 166 | |||
CR541953.1 | - | 2 | 166 | |||
D11428.1 | - | 3 | 354 | |||
DB466564.1 | - | 2 | 262 | |||
DB504971.1 | - | 3 | 405 | |||
HY119295.1 | - | 2 | 283 | |||
KR259963.1 | - | 3 | 373 | |||
KR259964.1 | - | 3 | 256 | |||
L03203.1 | - | 3 | 280 | |||
M94048.1 | - | 2 | 215 |