Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001329.2 | NP_001001329.1 | 5, 6 | 873 | ||
NM_001289102.1 | NP_001276031.1 | 4, 5 | 724 | |||
NM_001289103.1 | NP_001276032.1 | 5, 6 | 877 | |||
NM_001289104.1 | NP_001276033.1 | 5, 6 | 873 | |||
NM_002743.3 | NP_002734.2 | 5, 6 | 485 | |||
XM_011528130.1 | XP_011526432.1 | 6, 7 | 639 | |||
XM_011528131.1 | XP_011526433.1 | 6, 7 | 639 | |||
XM_011528132.1 | XP_011526434.1 | 6, 7 | 639 | |||
XM_017026977.1 | XP_016882466.1 | 5, 6 | 489 | |||
GenBank mRNA | AF144075.1 | - | 4, 5 | 346 | ||
AK130663.1 | - | 5, 6 | 501 | |||
AK225279.1 | - | 5, 6 | 499 | |||
AK225390.1 | - | 5, 6 | 481 | |||
AK290433.1 | - | 4, 5 | 719 | |||
AK296187.1 | - | 5, 6 | 500 | |||
BC013586.1 | - | 4, 5 | 283 | |||
BC128412.1 | - | 1, 2 | 85 | |||
BF984334.1 | - | 1, 2 | 37 | |||
J03075.1 | - | 5, 6 | 482 |