Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACCSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031854.2 | NP_001027025.2 | 8 | 1077 | ||
XM_017017714.1 | XP_016873203.1 | 11 | 1450 | |||
XM_017017715.1 | XP_016873204.1 | 8 | 1090 | |||
GenBank mRNA | AB231732.1 | - | 8 | 656 | ||
AB231733.1 | - | 5 | 384 | |||
AB231734.1 | - | 8 | 1077 | |||
BC144554.1 | - | 8 | 1090 |