Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000359.2 | NP_000350.1 | 8 | 1367 | ||
GenBank mRNA | AK291350.1 | - | 8 | 1334 | ||
AK299436.1 | - | 8 | 1136 | |||
AK315819.1 | - | 8 | 1367 | |||
AK315828.1 | - | 8 | 1327 | |||
AK315843.1 | - | 8 | 1334 | |||
BC034699.1 | - | 8 | 1318 | 4747045 | ||
D90287.1 | - | 8 | 1323 | |||
M55183.1 | - | 8 | 1296 | |||
M62925.1 | - | 8 | 1329 | |||
X57974.1 | - | 2 | 121 |