Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281463.1 | NP_001268392.1 | 7 | 1254 | ||
NM_006306.3 | NP_006297.2 | 6 | 1108 | |||
GenBank mRNA | AK291921.1 | - | 6 | 1067 | ||
BC064368.1 | - | 6 | 1039 | |||
BC080185.1 | - | 6 | 1043 | |||
BC112127.1 | - | 6 | 1048 | 8327595 | ||
BC171726.1 | - | 7 | 1194 | |||
BQ232418.1 | - | 4 | 758 | |||
BX640922.1 | - | 6 | 1151 | |||
D80000.2 | - | 6 | 980 | |||
S78271.1 | - | 6 | 1013 |