Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281463.1 | NP_001268392.1 | 5 | 734 | ||
NM_006306.3 | NP_006297.2 | 4 | 588 | |||
GenBank mRNA | AF176781.1 | - | 4 | 489 | ||
AK291921.1 | - | 4 | 547 | |||
BC064368.1 | - | 4 | 519 | |||
BC080185.1 | - | 4 | 523 | |||
BC112127.1 | - | 4 | 528 | 8327595 | ||
BC171726.1 | - | 5 | 674 | |||
BP312805.1 | - | 4 | 588 | |||
BQ232418.1 | - | 2 | 238 | |||
BX640922.1 | - | 4 | 631 | |||
D80000.2 | - | 4 | 460 | |||
S78271.1 | - | 4 | 493 |