Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003611.2 | NP_003602.1 | 21 | 3242 | ||
XM_005274604.2 | XP_005274661.1 | 20 | 3122 | |||
XM_005274606.3 | XP_005274663.1 | 22 | 3384 | |||
XM_011545592.2 | XP_011543894.1 | 22 | 3213 | |||
XM_011545594.2 | XP_011543896.1 | 20 | 2663 | |||
XM_011545595.1 | XP_011543897.1 | 20 | 2673 | |||
XM_011545597.1 | XP_011543899.1 | 21 | 3187 | |||
XM_011545598.1 | XP_011543900.1 | 10 | 1741 | |||
XM_017029909.1 | XP_016885398.1 | 21 | 2818 | |||
XM_017029910.1 | XP_016885399.1 | 22 | 3026 | |||
XM_017029911.1 | XP_016885400.1 | 13 | 2063 | |||
GenBank mRNA | AK225847.1 | - | 20 | 3049 | ||
AK290354.1 | - | 21 | 3224 | |||
AK297104.1 | - | 13 | 2022 | |||
BC096344.3 | - | 20 | 2937 | 40008704 | ||
BC096345.1 | - | 20 | 2951 | |||
CB134304.1 | - | 1 | 134 | |||
GU727634.1 | - | 20 | 2883 | |||
Y15164.1 | - | 21 | 3194 | |||
Y16355.1 | - | 21 | 3858 |