Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PQBP1 SIRNA Assays ›
Gene Symbol:
PQBP1
Gene Name:
polyglutamine binding protein 1
Gene Aliases:
MRX2, MRX55, MRXS3, MRXS8, NPW38, RENS1, SHS
Chromosome Location:
Chr.X: 48897862 - 48903145 on Build GRCh38
UniGene ID:
Hs.534384
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s527196
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PQBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001032381.1
NP_001027553.1
2
179
NM_001032382.1
NP_001027554.1
2
247
NM_001032383.1
NP_001027555.1
2
258
NM_001032384.1
NP_001027556.1
2
167
NM_001167989.1
NP_001161461.1
2
179
NM_001167990.1
NP_001161462.1
2
161
NM_001167992.1
NP_001161464.1
1
76
NM_005710.2
NP_005701.1
1
312
NM_144495.2
NP_652766.1
2
247
XM_005272571.3
XP_005272628.1
2
242
XM_005272572.4
XP_005272629.1
2
170
XM_011543884.2
XP_011542186.1
2
317
XM_017029207.1
XP_016884696.1
2
308
GenBank mRNA
AB016533.1
-
2
228
AB041833.1
-
1
58
AB041834.1
-
1
58
AB041835.1
-
1
58
AB041836.1
-
1
58
AJ005893.1
-
1
315
AJ242829.1
-
2
118
AJ973593.1
-
1
73
AJ973594.1
-
1
73
AJ973595.1
-
1
73
AJ973596.1
-
73
AJ973597.1
-
1
73
AJ973598.1
-
1
73
AJ973599.1
-
1
73
AJ973600.1
-
1
73
AJ973601.1
-
1
73
AJ973602.1
-
1
73
AJ973603.1
-
1
73
AJ973604.1
-
1
77
AJ973605.1
-
1
73
AJ973606.1
-
1
73
AJ973607.1
-
1
73
BC012358.1
-
2
139
4662649
BE396796.1
-
2
159
BI818141.1
-
2
141
BX362311.2
-
2
216
BX369697.2
-
2
399
CD051018.1
-
2
182
CN292761.1
-
2
247
CN292769.1
-
2
165
KU178500.1
-
1
58
KU178501.1
-
1
58
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
alternative mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
neuron projection development
regulation of RNA splicing
regulation of dendrite morphogenesis
cilium morphogenesis
DNA binding
transcription coactivator activity
protein binding
protein C-terminus binding
ribonucleoprotein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline