Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015139.2 | NP_055954.1 | 11 | 1295 | ||
XM_006710478.1 | XP_006710541.1 | 12 | 1376 | |||
XM_011541070.1 | XP_011539372.1 | 12 | 1376 | |||
GenBank mRNA | AB044343.1 | - | 11 | 943 | ||
AK289800.1 | - | 11 | 1293 | |||
AK296449.1 | - | 10 | 766 | |||
AK316023.1 | - | 5 | 288 | |||
BC093786.1 | - | 11 | 937 | 7939631 | ||
BC112031.1 | - | 11 | 949 | 8327499 | ||
D87449.1 | - | 11 | 997 |