Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199107.1 | NP_001186036.1 | 6 | 1384 | ||
NM_020705.2 | NP_065756.1 | 5 | 1366 | |||
XM_017023493.1 | XP_016878982.1 | 6 | 1426 | |||
XM_017023494.1 | XP_016878983.1 | 5 | 1496 | |||
XM_017023495.1 | XP_016878984.1 | 5 | 1408 | |||
GenBank mRNA | AB032997.1 | - | 6 | 1377 | ||
AB449911.1 | - | 5 | 1244 | |||
BC112389.1 | - | 5 | 1366 | 5502314 | ||
BC127014.1 | - | 4 | 1280 | 40118225 | ||
BC127015.1 | - | 5 | 1298 | 40118227 | ||
BX648283.1 | - | 2 | 113 |